Learn More
Beckman Coulter FisherbrandTM Petri Dishes, 100 x 15 mm, Sterile, 500/Pk
Catalog No. NC2306285
Encompass_Preferred
The FisherbrandTM NC2306285 is a polypropylene microcentrifuge tube rack that holds 0.5mL tubes. It has a 96-well format with alpha-numeric grid reference for easy sample tracking. Made from autoclavable polypropylene, this microtube rack is ideal for sample storage in freezers, refrigerators, and liquid nitrogen. It measures 127x86x45mm (LxWxH). With its rigid construction, it provides a stable base for microtubes. The rack allows easy retrieval while preventing accidental tipping.
Catalog No. NC2306285 Supplier Beckman Coulter Supplier No. 355650
The FisherbrandTM NC2306285 is a polypropylene microcentrifuge tube rack that holds 0.5mL tubes. It has a 96-well format with alpha-numeric grid reference for easy sample tracking. Made from autoclavable polypropylene, this microtube rack is ideal for sample storage in freezers, refrigerators, and liquid nitrogen. It measures 127x86x45mm (LxWxH). With its rigid construction, it provides a stable base for microtubes. The rack allows easy retrieval while preventing accidental tipping.