Learn More
Boston Bioproducts Inc Separating (Resolving) Buffer - 500ml
Catalog No. NC0228390
Encompass
The Separating (Resolving) Buffer is a critical component in SDS-Polyacrylamide Gel Electrophoresis (PAGE), a laboratory technique that is used to separate protein molecules based on molecular weight. This buffer maintains the pH of the separating or resolving gel at 8.8, which is a higher pH level than that of the stacking gel.
Catalog No. NC0228390 Supplier Boston Bioproducts Inc Supplier No. BP90500ML
The Separating (Resolving) Buffer is a critical component in SDS-Polyacrylamide Gel Electrophoresis (PAGE), a laboratory technique that is used to separate protein molecules based on molecular weight. This buffer maintains the pH of the separating or resolving gel at 8.8, which is a higher pH level than that of the stacking gel.