Learn More
JDM LAB SOLUTIONS LLC 500 ML STAINLESS STEEL DEWAR
Catalog No. NC1875557
Encompass
Product Features:Temperature range -190°C-200°C / Pharmaceutical grade stainless steel / Complete with vented stainless steel lids / Transports low-temperature gasses or high-temperature liquids
Catalog No. NC1875557 Supplier JDM LAB SOLUTIONS LLC Supplier No. SDF500
Product Features:Temperature range -190°C-200°C / Pharmaceutical grade stainless steel / Complete with vented stainless steel lids / Transports low-temperature gasses or high-temperature liquids