Learn More
VEOLIA WTS 500 RL System Suitability Set—Super Ios
Catalog No. NC9852907
Encompass
Set Includes (1) Reagent Water Blank - Rw, (1) Standard Solution - Rs(500 μg C/L as USP Sucrose), (1) Standard Solution - Rss (500 μg C/L asUSP 1,4-Benzoquinone), and (1) Rinse Water Standard. Super iOS standardsare packaged in 40 mL glass vials, inserted in a vial cartridge foruse with the Sievers Super iOS System.
Catalog No. NC9852907 Supplier VEOLIA WTS Supplier No. STD 74503
Set Includes (1) Reagent Water Blank - Rw, (1) Standard Solution - Rs(500 μg C/L as USP Sucrose), (1) Standard Solution - Rss (500 μg C/L asUSP 1,4-Benzoquinone), and (1) Rinse Water Standard. Super iOS standardsare packaged in 40 mL glass vials, inserted in a vial cartridge foruse with the Sievers Super iOS System.