Learn More
Ace Glass, Inc. 10mm 58cm stainless steel stir shaft, polished, removable button type, 28cm flat section
SDP
Catalog No. 509558207
Encompass
10mm O.D. 58cm stainless steel polished stir shaft, drilled type, with removable button. Stir Shaft has a 28cm flat section for use with agitators secured by set screws. For use with Trubore and PTFE bearings.
Catalog No. 50-955-8207 Supplier Ace Glass, Inc. Supplier No. 807407
10mm O.D. 58cm stainless steel polished stir shaft, drilled type, with removable button. Stir Shaft has a 28cm flat section for use with agitators secured by set screws. For use with Trubore and PTFE bearings.