Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ 5000 Series Disposable Half-Facepiece Respirators

Catalog No. 17674A Shop All 3M Products
Change view
Click to view available options
Color:
Black
Yellow
Size:
Small
Medium
Large
For Use With (Application):
Assembly, cleaning, grinding, machine operation, painting, welding
Petrochemical, chemical manufacturing, construction, transportation and electronics
Spray painting, pesticides, agriculture, construction and transportation
Includes:
Organic vapor cartridge, reusable storage bag
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size For Use With (Application) Includes
17-674A Black Small Petrochemical, chemical manufacturing, construction, transportation and electronics Organic vapor cartridge, reusable storage bag
17-674B Black Medium Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
17-674C Black Large Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
17-676A Yellow Small Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
17-676B Yellow Medium Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
17-676C Yellow Large Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
19-003-453B Black Medium Spray painting, pesticides, agriculture, construction and transportation Organic vapor cartridge, reusable storage bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. 17-674A Supplier 3M™ Supplier No. 7000126024
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Includes

Organic vapor cartridge and reusable storage bag.

Preassembled respirators are maintenance-free and require no spare parts

  • Ideally suited for intermittent use, plant shutdowns, and applications where respirators wear out or become difficult to clean in short periods of time
  • Color coded for easy size identification
  • Can be used with NIOSH-approved 3M filters N95, P95, and P100 for oil and non-oil particles
  • For convenience and simplicity, some respirator assemblies are packaged with 3M Filters P95 or P100 along with 3M Filter Retainer 501 or Filter Adapter 502
  • Some styles feature acid gas and/or organic vapor protection

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Compliance

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

TRUSTED_SUSTAINABILITY

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Black
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small
Facepiece Material Thermoplastic Elastomer
For Use With (Application) Petrochemical, chemical manufacturing, construction, transportation and electronics
For Use With (Equipment) 3M filters N95, P95, and P100 for oil and non-oil particles
Includes Organic vapor cartridge, reusable storage bag
Model 5101
Suspension Type 4-point
Weight (English) 1 lb.
Weight (Metric) 0.45 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.