Learn More
BURKLE INC close-it pharma maxi seal
Supplier:  BURKLE INC 53033018
Encompass_Preferred
Taking samples from sacks boxes bags etc with sampling lances is a common and time-proven practice This produces holes through which easily flowing bulk goods could escape from the container With the close-it seal this can be prevented in an uncomplicated manner close-it has been especially developed for this purpose The label has a sandwich-type structure (aluminium/paper) and seals the container hermetically (vapour barrier) This virtually prevents hygroscopic powders from clumping together The labels are available in different colours With printing in red blue green yellow black or neutral (without printing in white) close-it can be written on with ball-point pen pencil or felt-tip marker
Catalog No. 50-000-25972
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.