Learn More
Genscript Corporation HEK293/ACE2 Stable Cell Line
Supplier: Genscript Corporation M00770
Recombinant HEK293 cells stably express Angiotensin-converting enzyme 2 (ACE2) on the cell surface The surface expression of ACE2 is validated by FACS analysis This stable cell line product is designed for cell-based binding assays that measure the binding affinity and stability of antibody based biologics binding with ACE2 and can be utilized for SARS-Cov-2 pseudovirus neutralization assay for screening neutralizing antibodies