Learn More
Inorganic Ventures 5 Cesium Ionization Buffer
SDP
Catalog No. 50723540
Encompass_Preferred
Inorganic Ventures' 5% Cesium Ionization Buffer for ICP is a 500mL reference material set in a nitric acid matrix for stability. This cesium buffer is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-540 Supplier Inorganic Ventures Supplier No. CSNISB5500ML
Inorganic Ventures' 5% Cesium Ionization Buffer for ICP is a 500mL reference material set in a nitric acid matrix for stability. This cesium buffer is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search