Learn More
Medchemexpress LLC PD 128907 hydrochloride | 112960-16-4 | 99.5% | 285.77 g/mol | C14H20ClNO3 | 5 MG
SDP
Catalog No. 5000214888
Encompass_Preferred
PD 128907 hydrochloride is the hydrochloride salt of a potent, selective dopamine D3 receptor agonist used as a biochemical reagent for pharmacology and dopaminergic signaling research. Supplied as a solid, white to light yellow powder, it has reported purity of 99.51%, molecular formula C14H20ClNO3, and molecular weight 285.77 g/mol.
  • Potent D3 receptor agonist with EC50 of 0.64 nM.
  • Approximately 53-fold selectivity for D3 over D2 receptors.
  • High reported purity suitable for biochemical assays.
  • Solid form simplifies storage and handling.
  • Stable under recommended sealed storage conditions for short-term and long-term use.
Catalog No. 50-002-14888 Supplier Medchemexpress LLC Supplier No. HY100539A5MG
PD 128907 hydrochloride is the hydrochloride salt of a potent, selective dopamine D3 receptor agonist used as a biochemical reagent for pharmacology and dopaminergic signaling research. Supplied as a solid, white to light yellow powder, it has reported purity of 99.51%, molecular formula C14H20ClNO3, and molecular weight 285.77 g/mol.
  • Potent D3 receptor agonist with EC50 of 0.64 nM.
  • Approximately 53-fold selectivity for D3 over D2 receptors.
  • High reported purity suitable for biochemical assays.
  • Solid form simplifies storage and handling.
  • Stable under recommended sealed storage conditions for short-term and long-term use.