Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hispidol ((Z)-hispidol) | 5786-54-9 | 98.3% | 254.24 g·mol⁻¹ | C15H10O4 | 200 MG
SDP

Catalog No. 5000247435
Encompass_Preferred

Hispidol ((Z)-Hispidol) is a small-molecule TNF-α inhibitor used in research applications. It inhibits TNF-α-induced adhesion of monocytes to colon epithelial cells (IC50 = 0.50 μM) and is supplied as a solid analytical standard (CAS 5786-54-9).

  • Inhibits TNF-α-induced adhesion of monocytes to colon epithelial cells (IC50 = 0.50 μM).
  • Chemical formula C15H10O4.
  • Molecular weight 254.24 g·mol⁻¹.
  • Purity 98.3% (manufacturer listed).
  • Appearance solid, light yellow to yellow.
  • Suitable for research and analytical applications.
  • Storage: powder -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-47435 Supplier Medchemexpress LLC Supplier No. HY102040200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hispidol ((Z)-Hispidol) is a small-molecule TNF-α inhibitor used in research applications. It inhibits TNF-α-induced adhesion of monocytes to colon epithelial cells (IC50 = 0.50 μM) and is supplied as a solid analytical standard (CAS 5786-54-9).

  • Inhibits TNF-α-induced adhesion of monocytes to colon epithelial cells (IC50 = 0.50 μM).
  • Chemical formula C15H10O4.
  • Molecular weight 254.24 g·mol⁻¹.
  • Purity 98.3% (manufacturer listed).
  • Appearance solid, light yellow to yellow.
  • Suitable for research and analytical applications.
  • Storage: powder -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.