Learn More
Medchemexpress LLC Propargyl-PEG12-OH 250mg | 1036204-61-1 | 584.69 | C27H52O13 | 250 MG
SDP
Catalog No. 5000307854
Encompass_Preferred
Propargyl-PEG12-OH is a PEG-based PROTAC linker can be used in the synthesis of PROTACs Propargyl-PEG12-OH is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-003-07854 Supplier Medchemexpress LLC Supplier No. HY117045250MG
Propargyl-PEG12-OH is a PEG-based PROTAC linker can be used in the synthesis of PROTACs Propargyl-PEG12-OH is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups