Learn More
Sino Biological Recombinant Mouse BMPRIB Protein (hFc Tag)
Supplier:  Sino Biological 50004M02H100
Encompass_Preferred
A DNA sequence encoding the mouse Bmpr1b (NP_031586.1) (Lys14-Lys126) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1121