Learn More
Medchemexpress LLC Lusutrombopag | 1110766-97-6 | 98.88% | 591.55 | 200 MG
SDP
Catalog No. 5000401539
Encompass_Preferred
Lusutrombopag is an orally bioavailable thrombopoietin (TPO) receptor agonist, used for the treatment of chronic liver disease. It acts selectively on the human TPO receptor and activates signal transduction pathways that promote the proliferation and differentiation of bone marrow cells into megakaryocytes, thereby increasing platelet levels. This product is for research use only.
  • Orally bioavailable TPO receptor agonist
  • Treats chronic liver disease
  • Selectively acts on human TPO receptor
  • Promotes proliferation and differentiation of bone marrow cells
  • Increases platelet levels
Catalog No. 50-004-01539 Supplier Medchemexpress LLC Supplier No. HY19883200MG
Lusutrombopag is an orally bioavailable thrombopoietin (TPO) receptor agonist, used for the treatment of chronic liver disease. It acts selectively on the human TPO receptor and activates signal transduction pathways that promote the proliferation and differentiation of bone marrow cells into megakaryocytes, thereby increasing platelet levels. This product is for research use only.
  • Orally bioavailable TPO receptor agonist
  • Treats chronic liver disease
  • Selectively acts on human TPO receptor
  • Promotes proliferation and differentiation of bone marrow cells
  • Increases platelet levels