Learn More
Kactus Recombinant Human CDCP1 Protein is expressed from HEK293 with His tag at the C-Terminus.It contains Phe30-Leu666.
Supplier:  Kactus CDCHM101100UG
Encompass_Preferred
Human CDCP1 Protein (CDC-HM101). Activity of this product has been validated in ELISA and SPR, for detailed validation results and experiment setup, use the supplier link to see the product page detail. Endotoxin is less than 1 EU per μg by the LAL method. The purity of this protein is determined to be > 95% by Bis-Tris PAGE and > 95% by HPLC.
Catalog No. 50-004-62403