Learn More
Sino Biological Recombinant Anti-CD166/ALCAM Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 50005R311F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD166 / ALCAM (rM CD166 / ALCAM; Catalog#50005-M08H; NP_033785.1; Met1-Lys527) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-202-2783