Learn More
Gold Biotechnology Inc Leupeptin Hemisulfate 25 mg
SDP
Supplier:  Gold Biotechnology Inc L01025
Encompass_Preferred
Leupeptin hemisulfate (Ac-Leu-Leu-argininal) is a reversible inhibitor of serine and cysteine endopeptidases Leupeptin hemisulfate is capable of inhibiting plasmin trypsin papain kallikrein calpain and cathepsin B Leupeptin is usually used in a concentration range of 10 to 100 M Leupeptin is widely used in many protease inhibitor cocktails due to its potency low toxicity and range of inhibition Leupeptin is also used in a quantitative assay to measure macroautophagic flux in mammals and is being investigated for its use in treating malaria
Catalog No. 50-005-62029