Learn More
Waters Corp Sample Loop, 5 mL, Stainless Steel
Catalog No. 50792754 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This sample loop is designed for the precise introduction of samples into analytical systems. Crafted from robust stainless steel, it ensures reliability and compatibility within various chromatographic applications.
  • Capacity: 5 mL
  • Material: Stainless steel
Catalog No. 50-792-754 Supplier Waters Corp Supplier No. 700002449
This sample loop is designed for the precise introduction of samples into analytical systems. Crafted from robust stainless steel, it ensures reliability and compatibility within various chromatographic applications.
  • Capacity: 5 mL
  • Material: Stainless steel