Learn More
Ace Glass, Inc. Separatory funnel, 5000mL, 45/50, 8mm bore glass stopcock
SDP
Catalog No. 50974675
Encompass
5000mL separatory, globe type funnel, with neck and section between globe and stopcock shaped to provide a secure grip, for shaking contents or transporting. Funnel features a 45/50 standard taper top joint, and a 8mm bore glass stopcock plug.
Catalog No. 50-974-675 Supplier Ace Glass, Inc. Supplier No. 722428
5000mL separatory, globe type funnel, with neck and section between globe and stopcock shaped to provide a secure grip, for shaking contents or transporting. Funnel features a 45/50 standard taper top joint, and a 8mm bore glass stopcock plug.