Learn More
Chemglass Life Sciences Chemglass Glas-Col Series M 5000 mL Spherical Heating Mantle, 600 W (CG-11000-11)
SDP
Catalog No. NC1934297
Encompass_Preferred
The Chemglass Glas-Col Series M 5000 mL spherical heating mantle provides controlled bottom-only heating for 5,000 mL round-bottom flasks. With 600 W power and an internal operating temperature up to 450 °C, it is intended for laboratory heating applications and must be used with an external temperature controller and thermocouple or RTD.
  • 5,000 mL capacity for spherical flasks.
  • 600 W heating power.
  • Internal operating temperature up to 450 °C.
  • Hemispherical design covers only the bottom half of the flask for visual access.
  • Aluminum outer housing with glass-fabric heating element.
  • 115 V operation with a detachable 4 ft, 3-wire cord.
  • Requires external temperature controller and thermocouple or RTD; not intended for standalone temperature control.
Catalog No. NC1934297 Supplier Chemglass Life Sciences Supplier No. CG1100011
The Chemglass Glas-Col Series M 5000 mL spherical heating mantle provides controlled bottom-only heating for 5,000 mL round-bottom flasks. With 600 W power and an internal operating temperature up to 450 °C, it is intended for laboratory heating applications and must be used with an external temperature controller and thermocouple or RTD.
  • 5,000 mL capacity for spherical flasks.
  • 600 W heating power.
  • Internal operating temperature up to 450 °C.
  • Hemispherical design covers only the bottom half of the flask for visual access.
  • Aluminum outer housing with glass-fabric heating element.
  • 115 V operation with a detachable 4 ft, 3-wire cord.
  • Requires external temperature controller and thermocouple or RTD; not intended for standalone temperature control.