Learn More
Chemglass Life Sciences Flask, Round Bottom, 5000mL, Heavy Wall, Vertical 4-Neck, 45/50 CN, 29/42 SN's
SDP
Catalog No. CG153016
Encompass_Preferred
Flask, Round Bottom, 5000mL, Heavy Wall, 45/50 - 29/42, 4-Neck, Vertical. Round bottom, HEAVY WALL, four neck flask with standard taper outer joints on the center and vertical side necks.
Catalog No. CG153016 Supplier Chemglass Life Sciences Supplier No. CG153016
Flask, Round Bottom, 5000mL, Heavy Wall, 45/50 - 29/42, 4-Neck, Vertical. Round bottom, HEAVY WALL, four neck flask with standard taper outer joints on the center and vertical side necks.