Learn More
Chemglass Life Sciences 5000mL Reservoir, Chromatography, 45/50 Lower Rodaviss Inner
SDP
Catalog No. CG119411
Encompass_Preferred
5000mL Reservoir, Chromatography, 45/50 Lower Rodaviss Inner. Reservoir for use with CG-1192 and CG-1193 columns. Reservoir has a top 24/40 standard taper outer joint for use with CG-1195 flow controller, and the listed size lower inner joint. Supplied complete with one CG-182 open top cap, one CG-305 Viton o-ring and one CG-184 loosening ring.
Catalog No. CG119411 Supplier Chemglass Life Sciences Supplier No. CG119411
5000mL Reservoir, Chromatography, 45/50 Lower Rodaviss Inner. Reservoir for use with CG-1192 and CG-1193 columns. Reservoir has a top 24/40 standard taper outer joint for use with CG-1195 flow controller, and the listed size lower inner joint. Supplied complete with one CG-182 open top cap, one CG-305 Viton o-ring and one CG-184 loosening ring.