Learn More
Ace Glass, Inc. Fabric heating mantle that covers top and bottom of 1, 2 or3 neck 5000mL spherical flask. Lower half wattage is 600w at115 volts. Upper half wattage is 300w at 115 volts d, .
SDP
Catalog No. 50980640
Encompass
Catalog No. 50-980-640 Supplier Ace Glass, Inc. Supplier No. 1203125