Learn More
Chemglass Life Sciences 5000mL Single Neck Round Bottom Flask (RBF) , 24/40 Outer Joint
SDP
Catalog No. CG150629
Encompass_Preferred
5000mL Single Neck Round Bottomed Flask, 24/40 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.
Catalog No. CG150629 Supplier Chemglass Life Sciences Supplier No. CG150629
5000mL Single Neck Round Bottomed Flask, 24/40 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.