Learn More
Aqua Solutions Conductivity Standard 5,000 umhos @25°C NIST Traceable | 7447-40-7 | MFCD00011360 | 250mL
SDP
Catalog No. NC1749443
Encompass_Preferred
Conductivity Standard 5,000 umhos @25°C NIST Traceable | MW: 74.551 | 7447-40-7 | MFCD00011360 | 250mL
Catalog No. NC1749443 Supplier Aqua Solutions Supplier No. 6935250ML
Conductivity Standard 5,000 umhos @25°C NIST Traceable | MW: 74.551 | 7447-40-7 | MFCD00011360 | 250mL