Learn More
Sino Biological Recombinant Mouse PD-L1/B7-H1 Protein (ECD, hFc Tag), HPLC-verified
Catalog No. 501621131
Encompass_Preferred
A DNA sequence encoding the mouse Cd274 (NP_068693.1) (Met1-His239) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1131 Supplier Sino Biological Supplier No. 50010M02H100
A DNA sequence encoding the mouse Cd274 (NP_068693.1) (Met1-His239) was expressed with the Fc region of human IgG1 at the C-terminus.