Learn More
Sino Biological Recombinant Mouse BCL-xL/BCL2L1 Protein (aa 1-212, His Tag)
Supplier:  Sino Biological 50012M08E100
Encompass_Preferred
A DNA sequence encoding the mouse BCL2L1 (Q64373-1) (Met 1-Arg 212) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1138