Learn More
Sino Biological Recombinant Mouse B7-H4 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 50017M02H100
Encompass_Preferred
A DNA sequence encoding the mature form of mouse B7H4 (Q7TSP5) (phe 29-Ser 256) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1149