Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Labs Spectra/Por™ 7 3500 D MWCO Standard RC Pre-Treated Dialysis Kits
SDP

Catalog No. p-5001712 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
Diameter (Metric):
29 mm
34 mm
Length (Metric):
6.4 mL/cm vol./length
9.3 mL/cm vol./length
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
08-700-206 34 mm 9.3 mL/cm vol./length
08-700-205 29 mm 6.4 mL/cm vol./length
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 08-700-206 Supplier Spectrum™ Supplier No. 132112T
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

  • 1meter of pre-treated tubing, a pair of closures and 5 opening picks

With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds

  • Hydrophilic, symmetric porositiy
  • Equilibrated in water
  • Packaged wet in preservative solution of 0.05% sodium azide
  • For small experiments or qualifying a membrane for a particular application

Specifications

MWCO 3.5 kDa
Diameter (Metric) 34 mm
Length (Metric) 9.3 mL/cm vol./length
Quantity 1 m Tubing/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.