Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Labs Spectra/Por™ 2 and 4 12-14 kD MWCO Standard RC Dry Dialysis Kits
SDP

Catalog No. 08700160 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
Diameter (Metric):
20.4 mm
48 mm
Length (Metric):
18 mL/cm vol./length
3.3 mL/cm vol./length
Quantity:
5 m Tubing/Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric) Quantity
08-700-160 48 mm 18 mL/cm vol./length 5 m Tubing/Kit
08-700-158 20.4 mm 3.3 mL/cm vol./length 5 m Tubing/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-700-160 Supplier Spectrum™ Supplier No. 132709T
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • 1m or 5m of Spectra/Por 2 or 4 dialysis tubing
  • One Standard Spectra/Por Closure (for buoyance)
  • One weighted Spectra/Por Closure (for orientation)
  • Two adhesive tubing openers

For smaller dialysis experiments or evaluating dialysis tubing for dialysis application

  • Hydrophilic, symmetric porosity
  • Ensures higher permeability and basic dialysis
  • Available as dry with glycerin (soak in water)

Specifications

Product Type Dialysis Trial Kit
MWCO 12 to 14 kDa
Format Tubing
Diameter (Metric) 48 mm
Length (Metric) 18 mL/cm vol./length
Quantity 5 m Tubing/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.