Learn More
Sino Biological Recombinant Mouse CXADR/CAR Protein (His Tag)
Supplier:  Sino Biological 50019M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CXADR (NP_001020363.1) extracellular domain (Met 1-Gly 237) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1159