Learn More
Sino Biological Recombinant Mouse MCP-5/CCL12 Protein (His Tag)
Supplier:  Sino Biological 50021M08Y100
Encompass_Preferred
A DNA sequence encoding the mouse Ccl12 (NP_035461.2) (Gly23-Gly104) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1165