Learn More
Sino Biological Recombinant Mouse CD14 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50023M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 345) of mouse CD14 (NP_033971.1) precursor was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1169