Learn More
Sino Biological Recombinant Mouse Chemerin/RARRES2 Protein (His Tag)
Supplier:  Sino Biological 50024M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Rarres2 (NP_082128.1) (Met1-Ser156) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1174