Learn More
Sino Biological Recombinant Mouse CCL6 Protein (His Tag)
Supplier:  Sino Biological 50026M08H20
Encompass_Preferred
A DNA sequence encoding the mouse CCL6 (P27784) (Met 1-Ala 116) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1180