Learn More
Sino Biological Recombinant Mouse CD32B/Fcgr2b Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 50030M27HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Arg 217) of mouse FGGR2B (NP_001070657.1) as fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-162-1191