Learn More
Sino Biological Recombinant Mouse Erythropoietin R/EPOR Protein (His Tag)
Supplier:  Sino Biological 50031M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse EPOR (NP_034279.3) (Met 1-Pro 249) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1195