Learn More
Sino Biological Recombinant Mouse CD16-2/FCGR4/Fc gamma RIV Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50036M08H100
Encompass_Preferred
The amino acid sequence corronding to Met 1-Gln 203 of mouse FCGR4 (NP_653142.2) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1205