Learn More
Sino Biological Recombinant Mouse CD16-2/FCGR4/Fc gamma RIV Protein (His & AVI Tag)
Supplier:  Sino Biological 50036M27H100
Encompass_Preferred
The amino acid sequence corronding to (Met 1-Gln 203) of mouse FCGR4 (NP_653142.2) was fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus.
Catalog No. 50-162-1207