Learn More
Sino Biological Recombinant Mouse L-selectin/CD62L Protein (His Tag)
Supplier:  Sino Biological 50045M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Asn 332) of mouse SELL (NP_035476.1) precursor was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1230