Learn More
Sino Biological Recombinant Mouse TARC/CCL17 Protein
Supplier:  Sino Biological 50046MNAB50
Encompass_Preferred
A DNA sequence encoding the mouse CCL17 (AAD56601.1) (Met 1-Pro 93) was expressed and purified.
Catalog No. 50-162-1233