Learn More
Sino Biological Recombinant Mouse CCL20/MIP-3 Protein
Supplier:  Sino Biological 50048MNAE100
Encompass_Preferred
A DNA sequence encoding the mouse CCL20 (NP_058656.1) (Ala28-Met97) was expressed with an initial Met.
Catalog No. 50-162-1236