Learn More
Sino Biological Recombinant Mouse SFRP4 Protein (His Tag)
Supplier:  Sino Biological 50053M08H50
Encompass_Preferred
A DNA sequence encoding the mouse sFRP4 (NP_057896.1) (Met 1-Ser 351) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1245