Learn More
Sino Biological Recombinant Mouse M-CSFR/CSF1R Protein (His Tag)
Supplier:  Sino Biological 50059M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CSF1R (NP_001032948.2) extracellular domain (Met 1-Ser 511) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1253