Learn More
Sino Biological Recombinant Mouse Lipocalin-2/NGAL Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50060M08H100
Encompass_Preferred
A DNA sequence encoding the mouse LCN2 (NP_032517.1) precursor (Met 1-Asn 200) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1256