Learn More
Sino Biological Recombinant Mouse Lipocalin-2/NGAL Protein, HPLC-verified
Supplier:  Sino Biological 50060MNAH100
Encompass_Preferred
A DNA sequence encoding the mouse LCN2 (NP_032517.1) (Met1-Asn200) was expressed.
Catalog No. 50-162-1258