Learn More
Sino Biological Recombinant Mouse IL-13 R alpha 2/IL13RA2 Protein (His & hFc Tag)
Supplier:  Sino Biological 50061M03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Lys 334) of mouse IL13Rα2 (NP_032382.1) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1260