Learn More
Sino Biological Recombinant Mouse MD1 Protein (His Tag)
Supplier:  Sino Biological 50062M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 162) of mouse LY86 (NP_034875.1) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1264