Learn More
Sino Biological Recombinant Mouse LYVE1 Protein (hFc Tag)
Supplier:  Sino Biological 50065M02H50
Encompass_Preferred
A DNA sequence encoding the mouse LYVE1 (Q8BHC0) (Met1-Gly228) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1271