Learn More
Sino Biological Recombinant Mouse MIF Protein (His Tag)
Supplier:  Sino Biological 50066M08H50
Encompass_Preferred
A DNA sequence encoding the mouse MIF (P34884) precursor (Pro 2-Ala 115) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-162-1273